2,4-Dichloro-6-morpholino-1,3,5-triazine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(4,6-Dichloro-1,3,5-triazin-2-yl)morpholine, 2,4-Dichloro-6-(4-morpholinyl)-1,3,5-triazine
D4814-1G
D4814-5G
TCD4814-1GEA
111
USD
TCD4814-1G
TCD4814-5G
2,4-Dichloro-6-morpholino-1,3,5-triazine ≥98.0% (by GC)
2,4-Dichloro-6-morpholino-1,3,5-triazine
Formula:
C₇H₈Cl₂N₄O MW: 235.0706 g/mol |
MDL Number:
MFCD00460178 CAS Number: 6601-22-5 EINECS: 229-544-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...