3-Methyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Formyl-3-methylpyrazole, 3-Methylpyrazole-4-carboxaldehyde
M2643-1G
M2643-200MG
TCM2643-1GEA
159
USD
TCM2643-1G
TCM2643-200MG
3-Methyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
3-Methyl-1H-pyrazole-4-carbaldehyde
Formula:
C5H6N2O MW: 110.12 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01313822 CAS Number: 112758-40-4 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...