4-Amino-5,6-dichloropyrimidine
Supplier: TCI America
|
Warning
|
A3058-25G
A3058-5G
TCA3058-25GEA
172
USD
TCA3058-25G
TCA3058-5G
4-Amino-5,6-dichloropyrimidine
4-Amino-5,6-dichloropyrimidine
Formula:
C₄H₃Cl₂N₃ |
MDL Number:
MFCD09837293 CAS Number: 310400-38-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...