6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥94.0% (by GC)

Supplier: TCI America
Warning

B4402-1G B4402-5G
TCB4402-1GEA 64 USD
TCB4402-1G TCB4402-5G
6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥94.0% (by GC)
6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Formula: C₈H₆BrNO₂
MDL Number: MFCD00461173
CAS Number: 24036-52-0

Order Now

Specification Test Results

Purity (GC) min. 94.0 %
Melting point 219.0 to 223.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR