2-Amino-5-bromobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A3063-1G
A3063-200MG
TCA3063-1GEA
143
USD
TCA3063-1G
TCA3063-200MG
2-Amino-5-bromobenzaldehyde ≥98.0% (by GC)
2-Amino-5-bromobenzaldehyde
Formula:
C₇H₆BrNO Storage Temperature: Refrigerator |
MDL Number:
MFCD10696879 CAS Number: 29124-57-0 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Acid Value | max. 2.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...