5-Chloropicolinonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 5-Chloro-2-cyanopyridine

C3023-25G C3023-5G
TCC3023-25GEA 409 USD
TCC3023-25G TCC3023-5G
5-Chloropicolinonitrile ≥98.0% (by GC)
5-Chloropicolinonitrile
Formula: C6H3ClN2
MW: 138.55 g/mol
Boiling Pt: 110°C
MDL Number: MFCD03788835
CAS Number: 89809-64-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR