5-(Hydroxymethyl)furan-2-carboxylic Acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Hydroxymethyl-2-furancarboxylic acid, Sumiki's acid, 5-(Hydroxymethyl)-2-furoic acid

H1750-1G H1750-5G
TCH1750-1GEA 40 USD
TCH1750-1G TCH1750-5G
5-(Hydroxymethyl)furan-2-carboxylic Acid ≥98.0% (by GC, titration analysis)
5-(Hydroxymethyl)furan-2-carboxylic Acid
Formula: C6H6O4
MW: 142.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03274472
CAS Number: 6338-41-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR