2,4,6-Trichloropyrimidine-5-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

T3100-1G T3100-5G
TCT3100-1GEA 39 USD
TCT3100-1G TCT3100-5G
2,4,6-Trichloropyrimidine-5-carbaldehyde ≥98.0% (by GC)
2,4,6-Trichloropyrimidine-5-carbaldehyde
Formula: C₅HCl₃N₂O
MDL Number: MFCD02257700
CAS Number: 50270-27-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR