2,4-Dichloronicotinc acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,4-Dichloropyridine-3-carboxylic acid, 2,4-Dichloro-3-pyridinecarboxylic acid, 2,4-Dichloronicotinic acid

D4901-1G D4901-5G
TCD4901-1GEA 34 USD
TCD4901-1G TCD4901-5G
2,4-Dichloronicotinc acid ≥98.0% (by GC, titration analysis)
2,4-Dichloronicotinc acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD06796292
CAS Number: 262423-77-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR