1-Boc-4-(aminomethyl)piperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4445-1G
B4445-5G
TCB4445-1GEA
69
USD
TCB4445-1G
TCB4445-5G
1-Boc-4-(aminomethyl)piperidine ≥98.0% (by GC, titration analysis)
1-Boc-4-(aminomethyl)piperidine
Formula:
C₁₁H₂₂N₂O₂ Flash Pt: 110°C |
MDL Number:
MFCD01076207 CAS Number: 144222-22-0 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...