6,7-Dimethoxy-4-hydroxyquinoline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
D4695-1G
D4695-5G
TCD4695-1GEA
155
USD
TCD4695-1G
TCD4695-5G
6,7-Dimethoxy-4-hydroxyquinoline ≥98.0% (by HPLC, titration analysis)
6,7-Dimethoxy-4-hydroxyquinoline
Formula:
C₁₁H₁₁NO₃ |
MDL Number:
MFCD07368026 CAS Number: 13425-93-9 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 227.0 to 231.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...