2-(Trifluoromethyl)isonicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T3269-1G T3269-200MG
76220-540EA 50 USD
76220-540 76220-542
2-(Trifluoromethyl)isonicotinic acid ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)isonicotinic acid
Formula: C₇H₄F₃NO₂
MDL Number: MFCD07774132
CAS Number: 131747-41-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point gt. 219.0 and le 223.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR