Ethyl-3-oxo-3-(pyridin-4-yl)propanoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E1032-1G E1032-5G
TCE1032-1GEA 36 USD
TCE1032-1G TCE1032-5G
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate ≥98.0% (by GC, titration analysis)
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate
Formula: C₁₀H₁₁NO₃
Boiling Pt: 169°C
MDL Number: MFCD00094021
CAS Number: 26377-17-3
EINECS: 247-635-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR