Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate, Ethyl 2-acetyl-3-(dimethylamino)acrylate

E1179-25G E1179-5G
TCE1179-25GEA 153 USD
TCE1179-25G TCE1179-5G
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95.0% (by GC, titration analysis)
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Boiling Pt: 150°C
MDL Number: MFCD00087563
CAS Number: 51145-57-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR