2-Methyl-5-nitrobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2957-1G
M2957-5G
TCM2957-1GEA
124
USD
TCM2957-1G
TCM2957-5G
2-Methyl-5-nitrobenzaldehyde ≥98.0% (by GC)
2-Methyl-5-nitrobenzaldehyde
Formula:
C₈H₇NO₃ Boiling Pt: 114°C |
MDL Number:
MFCD03840506 CAS Number: 16634-91-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...