tert-Butyl-4-formylpiperidine-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America
Warning

B3873-1G B3873-5G
TCB3873-1GEA 22 USD
TCB3873-1G TCB3873-5G
tert-Butyl-4-formylpiperidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-4-formylpiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Melting point 32.0 to 37.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR