tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: 1-Boc-4-(tosyloxymethyl)piperidine, 1-(tert-Butoxycarbonyl)-4-(tosyloxymethyl)piperidine, tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-[(p-toluenesulphonyloxy)methyl]piperidine

B4759-25G B4759-5G
TCB4759-25GEA 250 USD
TCB4759-25G TCB4759-5G
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate ≥98.0% (by total nitrogen basis)
tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate
Formula: C₁₈H₂₇NO₅S
MW: 369.48 g/mol
MDL Number: MFCD05864740
CAS Number: 166815-96-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR