5-Bromo-1H-indole-3-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

B4840-1G B4840-5G
TCB4840-1GEA 26 USD
TCB4840-1G TCB4840-5G
5-Bromo-1H-indole-3-carbaldehyde ≥98.0% (by GC)
5-Bromo-1H-indole-3-carbaldehyde
Formula: C₉H₆BrNO
MDL Number: MFCD00152016
CAS Number: 877-03-2
EINECS: 212-884-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 204.0 to 208.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR