2,4-Di-tert-pentylphenol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5320-25G D5320-400G
76219-908EA 18 USD
76219-908 76219-910
2,4-Di-tert-pentylphenol ≥98.0% (by GC)
2,4-Di-tert-pentylphenol
Formula: C₁₆H₂₆O
Boiling Pt: 143°C
MDL Number: MFCD00041929
CAS Number: 120-95-6
EINECS: 204-439-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White or Colorless to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR