1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid

C2954-25G C2954-5G
TCC2954-25GEA 187.3 USD
TCC2954-25G TCC2954-5G
1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉ClO₂
MW: 196.64 g/mol
MDL Number: MFCD00001289
CAS Number: 72934-37-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR