2-Bromo-4,6-dimethylpyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B5254-1G B5254-5G
TCB5254-1GEA 129 USD
TCB5254-1G TCB5254-5G
2-Bromo-4,6-dimethylpyridine ≥98.0% (by titrimetric analysis)
2-Bromo-4,6-dimethylpyridine
Formula: C₇H₈BrN
Boiling Pt: 68°C
MDL Number: MFCD00082591
CAS Number: 4926-26-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR