3-Acetyl-2-chloropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

A2795-1G A2795-5G
TCA2795-1GEA 62.8 USD
TCA2795-1G TCA2795-5G
3-Acetyl-2-chloropyridine ≥98.0% (by GC)
3-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
Boiling Pt: 114°C
Flash Pt: 110°C
MDL Number: MFCD03840751
CAS Number: 55676-21-6

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR