4-(Tetrahydropyran-2-yloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid
T3325-25G
T3325-5G
TCT3325-25GEA
232
USD
TCT3325-25G
TCT3325-5G
4-(Tetrahydropyran-2-yloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Tetrahydropyran-2-yloxy)phenylboronic acid
Formula:
C₁₁H₁₅BO₄ MW: 222.05 g/mol Melting Pt: 142°C |
MDL Number:
MFCD02183525 CAS Number: 182281-01-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 98.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...