4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester

T3477-1G T3477-5G
TCT3477-1GEA 62 USD
TCT3477-1G TCT3477-5G
4-(Morpholinomethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Morpholinomethyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₃
MDL Number: MFCD04974052
CAS Number: 364794-79-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR