trans-4-(4-Vinylcyclohexyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

V0135-1G V0135-5G
TCV0135-1GEA 41.8 USD
TCV0135-1G TCV0135-5G
trans-4-(4-Vinylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Vinylcyclohexyl)benzonitrile
Formula: C₁₅H₁₇N
MDL Number: MFCD11053490
CAS Number: 96184-42-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR