trans-4-(4-Butylcyclohexyl)benzonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
B4924-25G
B4924-5G
TCB4924-25GEA
239.6
USD
TCB4924-25G
TCB4924-5G
trans-4-(4-Butylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Butylcyclohexyl)benzonitrile
Formula:
C₁₇H₂₃N |
MDL Number:
MFCD00799425 CAS Number: 61204-00-0 EINECS: 262-655-0 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...