4-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(tert-Butoxycarbonyl)benzeneboronic acid, (4-(tert-Butoxycarbonyl)phenyl)boronic acid, 4-tert-Butoxycarbonylphenylboronic acid
B4772-1G
B4772-200MG
B4772-5G
TCB4772-1GEA
91
USD
TCB4772-1G
TCB4772-200MG
TCB4772-5G
4-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(tert-Butoxycarbonyl)phenylboronic acid
Formula:
C₁₁H₁₅BO₄ MW: 222.05 g/mol |
MDL Number:
MFCD03411946 CAS Number: 850568-54-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...