3-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-(tert-Butoxycarbonyl)benzeneboronic acid, 3-tert-Butoxycarbonylphenylboronic acid

B4726-1G B4726-5G
TCB4726-1GEA 39 USD
TCB4726-1G TCB4726-5G
3-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(tert-Butoxycarbonyl)phenylboronic acid
Formula: C₁₁H₁₅BO₄
MW: 222.05 g/mol
Melting Pt: 102°C
MDL Number: MFCD01630855
CAS Number: 220210-56-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR