Methyl-6-aminopicolinate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester
M2865-1G
M2865-5G
TCM2865-1GEA
30
USD
TCM2865-1G
TCM2865-5G
Methyl-6-aminopicolinate ≥98.0% (by GC)
Methyl 6-aminopicolinate
Formula:
C₇H₈N₂O₂ MW: 152.1500 g/mol |
MDL Number:
MFCD00233712 CAS Number: 36052-26-3 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...