Methyl-5-bromopicolinate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

M2614-1G M2614-5G
TCM2614-1GEA 21 USD
TCM2614-1G TCM2614-5G
Methyl-5-bromopicolinate ≥98.0% (by GC)
Methyl 5-bromopicolinate
Formula: C₇H₆BrNO₂
MDL Number: MFCD04112493
CAS Number: 29682-15-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR