2-Amino-5-chloro-4-methylpyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2945-1G A2945-5G
TCA2945-1GEA 41 USD
TCA2945-1G TCA2945-5G
2-Amino-5-chloro-4-methylpyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-chloro-4-methylpyridine
Formula: C₆H₇ClN₂
MDL Number: MFCD06410759
CAS Number: 36936-27-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR