Methyl-3-methylthiophene-2-carboxylate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-methylthiophene-2-carboxylate

M2662-1G M2662-5G
TCM2662-1GEA 28 USD
TCM2662-1G TCM2662-5G
Methyl-3-methylthiophene-2-carboxylate ≥97.0% (by GC)
Methyl 3-methylthiophene-2-carboxylate
Formula: C₇H₈O₂S
MW: 156.20 g/mol
Boiling Pt: 217°C
MDL Number: MFCD00234332
CAS Number: 81452-54-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR