4-Cyano-4'-ethoxybiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

C3153-25G C3153-5G
TCC3153-25GEA 394.4 USD
TCC3153-25G TCC3153-5G
4-Cyano-4'-ethoxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-ethoxybiphenyl
Formula: C₁₅H₁₃NO
MDL Number: MFCD01218033
CAS Number: 58743-78-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 98.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR