5-Methoxypyridine-3-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid
M2810-1G
M2810-5G
TCM2810-1GEA
92
USD
TCM2810-1G
TCM2810-5G
5-Methoxypyridine-3-boronic acid (contains varying amounts of Anhydride)
5-Methoxypyridine-3-boronic acid
Formula:
C₆H₈BNO₃ |
MDL Number:
MFCD03425977 CAS Number: 850991-69-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...