Methyl-3-bromo-2-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2543-25G M2543-5G
TCM2543-25GEA 329 USD
TCM2543-25G TCM2543-5G
Methyl-3-bromo-2-methylbenzoate ≥98.0% (by GC)
Methyl 3-bromo-2-methylbenzoate
Formula: C₉H₉BrO₂
Boiling Pt: 140°C
MDL Number: MFCD09031772
CAS Number: 99548-54-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR