Methyl-2-bromo-3-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 2-bromo-3-methylbenzoate

M2546-25G M2546-5G
TCM2546-25GEA 153 USD
TCM2546-25G TCM2546-5G
Methyl-2-bromo-3-methylbenzoate ≥98.0% (by GC)
Methyl 2-bromo-3-methylbenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00144762
CAS Number: 131001-86-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR