Methyl-3-bromo-2-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 3-bromo-2-fluorobenzoate

M2711-1G M2711-5G
TCM2711-1GEA 46 USD
TCM2711-1G TCM2711-5G
Methyl-3-bromo-2-fluorobenzoate ≥98.0% (by GC)
Methyl 3-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD09261255
CAS Number: 206551-41-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR