2,6-Dichloroquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

D5429-1G D5429-5G
76219-976EA 64 USD
76219-976 76219-978
2,6-Dichloroquinoline ≥98.0% (by GC)
2,6-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.0500 g/mol
Boiling Pt: 172°C
MDL Number: MFCD03427194
CAS Number: 1810-72-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR