Methyl-2,3-diaminobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 2,3-diaminobenzoate

M2979-1G M2979-5G
TCM2979-1GEA 86 USD
TCM2979-1G TCM2979-5G
Methyl-2,3-diaminobenzoate ≥98.0% (by GC)
Methyl 2,3-diaminobenzoate
Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD04038589
CAS Number: 107582-20-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR