3-Phenyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Phenylisoxazole-5-carboxylic acid

P2390-1G P2390-5G
TCP2390-1GEA 68 USD
TCP2390-1G TCP2390-5G
3-Phenyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)
3-Phenyl-5-isoxazolecarboxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17
MDL Number: MFCD01936010
CAS Number: 14442-12-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR