4-Bromo-1H-indazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Bromoindazole
B4631-1G
B4631-5G
TCB4631-1GEA
56
USD
TCB4631-1G
TCB4631-5G
4-Bromo-1H-indazole ≥98.0% (by GC)
4-Bromo-1H-indazole
Formula:
C₇H₅BrN₂ MW: 197.04 g/mol |
MDL Number:
MFCD05664001 CAS Number: 186407-74-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...