3,4,6-Trichloropyridazine ≥97.0% (by GC)

Supplier: TCI America
Warning

T3417-1G T3417-5G
TCT3417-1GEA 24 USD
TCT3417-1G TCT3417-5G
3,4,6-Trichloropyridazine ≥97.0% (by GC)
3,4,6-Trichloropyridazine
Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
Boiling Pt: 98°C
MDL Number: MFCD00834957
CAS Number: 6082-66-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR