3-Amino-6-bromo-2-methylpyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

A2928-1G A2928-5G
TCA2928-1GEA 45 USD
TCA2928-1G TCA2928-5G
3-Amino-6-bromo-2-methylpyridine ≥97.0% (by GC, titration analysis)
3-Amino-6-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03095086
CAS Number: 126325-47-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR