6-Bromo-2-chloroquinoline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5628-1G
B5628-5G
TCB5628-1GEA
59
USD
TCB5628-1G
TCB5628-5G
6-Bromo-2-chloroquinoline ≥98.0% (by GC)
6-Bromo-2-chloroquinoline
Formula:
C₉H₅BrClN MW: 242.50 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04115272 CAS Number: 1810-71-5 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...