Methyl-2-iodo-5-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America

M3019-1G M3019-5G
TCM3019-1GEA 24 USD
TCM3019-1G TCM3019-5G
Methyl-2-iodo-5-methylbenzoate ≥98.0% (by GC)
Methyl 2-iodo-5-methylbenzoate
Formula: C₉H₉IO₂
Boiling Pt: 105°C
Storage Temperature: Refrigerator
MDL Number: MFCD12198129
CAS Number: 103440-52-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR