4-(Methylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-(Methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)phenylboronic acid

M3056-1G M3056-5G
TCM3056-1GEA 77 USD
TCM3056-1G TCM3056-5G
4-(Methylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Methylcarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₃
MDL Number: MFCD03788427
CAS Number: 121177-82-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR