2-Methyl-4-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methylthiazole-4-carboxylic acid

M2802-1G M2802-5G
TCM2802-1GEA 100 USD
TCM2802-1G TCM2802-5G
2-Methyl-4-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-4-thiazolecarboxylic acid
Formula: C₅H₅NO₂S
MW: 143.1637 g/mol
Melting Pt: 144°C
MDL Number: MFCD03407332
CAS Number: 35272-15-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR