2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98.0% (by GC)

Supplier: TCI America
Warning

C3208-25G C3208-5G
TCC3208-25GEA 433 USD
TCC3208-25G TCC3208-5G
2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98.0% (by GC)
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
MDL Number: MFCD01631469
CAS Number: 64372-62-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR