1-Dodecyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Dodecyl-3-methylimidazolium Bromide

D5356-25G D5356-5G
TCD5356-25GEA 360 USD
TCD5356-25G TCD5356-5G
1-Dodecyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
1-Dodecyl-3-methylimidazolium bromide
Formula: C₁₆H₃₁BrN₂
MW: 331.3347 g/mol
MDL Number: MFCD17014729
CAS Number: 61546-00-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR