(S)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A3309-1G A3309-250MG
TCA3309-1GEA 209 USD
TCA3309-1G TCA3309-250MG
(S)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
(S)-3-Aminobutyric acid
Formula: C₄H₉NO₂
MDL Number: MFCD00270347
CAS Number: 3775-72-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +34.0 to +38.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR